2-Methyl-5-pyridineboronic acid pinacol ester 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Methylpyridine-5-boronic acid pinacol ester, 6-Methylpyridine-3-boronic acid pinacol ester, 2-(6-Methyl-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 6-Methyl-3-pyridylboronic acid Pinacol ester

OR61383-10G OR61383-25G OR61383-5G
APOSOR61383-10GEA 1040 DKK
APOSOR61383-10G APOSOR61383-25G APOSOR61383-5G
2-Methyl-5-pyridineboronic acid pinacol ester 97%
2-Methyl-5-pyridineboronic acid pinacol ester
Formel: C₁₂H₁₈BNO₂
Molekylvægt: 219.0878 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD08669608
CAS-nummer: 610768-32-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR