N-Carbobenzoxy-L-proline 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Z-Pro-OH, Z-L-Prolin, N-Cbz-L-proline, N-Carbobenzyloxy-L-proline, N-Benzyloxycarbonyl-L-proline

OR61280-100G OR61280-25G OR61280-500G
APOSOR61280-100GEA 186 DKK
APOSOR61280-100G APOSOR61280-25G APOSOR61280-500G
N-Carbobenzoxy-L-proline 98%
N-Carbobenzoxy-L-proline
Formel: C₁₃H₁₅NO₄
Smeltepunkt: 76…78 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003170
CAS-nummer: 1148-11-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR