Glycintert-butylester 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: tert-Butyl 2-aminoacetate, tert-Butyl Glycinate

OR61270-25G OR61270-5G
APOSOR61270-25GEA 262 DKK
APOSOR61270-25G APOSOR61270-5G
Glycintert-butylester 97%
Glycintert-butylester
Formel: C₆H₁₃NO₂
Kogepunkt: 29…31 °C/2mmHg
Massefylde: 0,96 g/cm³ (25 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00038194
CAS-nummer: 6456-74-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR