2-Amino-5-bromothiazole hydrobromide 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Amino-5-bromothiazole monohydrobromide, 2-Amino-5-bromothiazole HBr

OR6124-25G OR6124-5G OR6124-1G
APOSOR6124-25GEA 1360 DKK
APOSOR6124-25G APOSOR6124-5G APOSOR6124-1G
2-Amino-5-bromothiazole hydrobromide 97%
2-Amino-5-bromothiazole hydrobromide
Formel: C₃H₃BrN₂S
Molekylvægt: 259.95 g/mol
Smeltepunkt: 163…165 °C (dec.)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00012712
CAS-nummer: 61296-22-8
EINECS: 262-699-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR