3-Brom-5-nitroanisol 98%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
1-Bromo-3-methoxy-5-nitrobenzene
OR61069-25G
OR61069-5G
APOSOR61069-25GEA
765
DKK
APOSOR61069-25G
APOSOR61069-5G
3-Brom-5-nitroanisol 98%
3-Brom-5-nitroanisol
Formel:
C₇H₆BrNO₃ Smeltepunkt: 85…88 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD04112752 CAS-nummer: 16618-67-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...