5-Bromo-2-carboxy-3-methylpyridine

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 5-Bromo-3-methylpyridine-2-carboxylic acid, 5-Bromo-3-methylpicolinic acid

OR61015-1G OR61015-5G
APOSOR61015-1GEA 164 DKK
APOSOR61015-1G APOSOR61015-5G
5-Bromo-2-carboxy-3-methylpyridine
5-Bromo-2-carboxy-3-methylpyridine
Formel: C₇H₆BrNO₂
Molekylvægt: 216.03 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD07375125
CAS-nummer: 886365-43-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR