3-Chlorisoquinolin 98%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
OR6098-1G
OR6098-5G
APOSOR6098-1GEA
426
DKK
APOSOR6098-1G
APOSOR6098-5G
3-Chlorisoquinolin 98%
3-Chlorisoquinolin
Formel:
C₉H₆ClN Molekylvægt: 163.6000 g/mol Smeltepunkt: 42…45 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD05982009 CAS-nummer: 19493-45-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...