(4-Bromophenyl)methanesulphonyl chloride 96%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 4-Bromobenzylsulphonyl chloride

OR6075-1G OR6075-500MG OR6075-5G
APOSOR6075-1GEA 393 DKK
APOSOR6075-1G APOSOR6075-500MG APOSOR6075-5G
(4-Bromophenyl)methanesulphonyl chloride 96%
(4-Bromophenyl)methanesulphonyl chloride
Formel: C₇H₆BrClO₂S
Molekylvægt: 269.5433 g/mol
Smeltepunkt: 117…121 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD04973454
CAS-nummer: 53531-69-4
UN nummer: 3261
ADR: 8 II

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR