(3-Bromo-2-pyridinyl)methanol
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR60200-1G
OR60200-5G
APOSOR60200-1GEA
183
DKK
APOSOR60200-1G
APOSOR60200-5G
(3-Bromo-2-pyridinyl)methanol
(3-Bromo-2-pyridinyl)methanol
Formel:
C₆H₆BrNO Smeltepunkt: 55…57 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD03095348 CAS-nummer: 52378-64-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...