Methyl-6-chloro-3-methylpicolinate

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Methyl 6-chloro-3-methylpicolinate, Methyl 6-chloro-3-methylpyridine-2-carboxylate

OR60156-1G OR60156-250MG
APOSOR60156-1GEA 414 DKK
APOSOR60156-1G APOSOR60156-250MG
Methyl-6-chloro-3-methylpicolinate
Methyl-6-chloro-3-methylpicolinate
Formel: C₈H₈ClNO₂
Opbevaringstemperatur: Køleskab
MDL Number: MFCD12756112
CAS-nummer: 878207-92-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR