6-Bromo-3-iodo-1H-indazole
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR60115-1G
OR60115-5G
OR60115-25G
APOSOR60115-1GEA
164
DKK
APOSOR60115-1G
APOSOR60115-5G
APOSOR60115-25G
6-Bromo-3-iodo-1H-indazole
6-Bromo-3-iodo-1H-indazole
Formel:
C₇H₄BrIN₂ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD07781599 CAS-nummer: 885521-88-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...