4-(4-Chlorobenzoyl)pyridine

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR59834-1G OR59834-5G
APOSOR59834-1GEA 741 DKK
APOSOR59834-1G APOSOR59834-5G
4-(4-Chlorobenzoyl)pyridine
4-(4-Chlorobenzoyl)pyridine
Formel: C₁₂H₈ClNO
Molekylvægt: 217.66 g/mol
Smeltepunkt: 107…109 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00006431
CAS-nummer: 14548-48-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR