(2,4-Dinitrophenyl)eddikesyre 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR5974-25G OR5974-5G
APOSOR5974-25GEA 1590 DKK
APOSOR5974-25G APOSOR5974-5G
(2,4-Dinitrophenyl)eddikesyre 98%
(2,4-Dinitrophenyl)eddikesyre
Formel: (C₆H₃(NO₂)₂)CH₂COOH
Smeltepunkt: 171…174 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007227
CAS-nummer: 643-43-6
EINECS: 211-398-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR