4-Nitrobenzamidine hydrochloride

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR59393-1G OR59393-250MG
APOSOR59393-1GEA 186 DKK
APOSOR59393-1G APOSOR59393-250MG
4-Nitrobenzamidine hydrochloride
4-Nitrobenzamidine hydrochloride
Formel: C₇H₇N₃O₂
Molekylvægt: 201.61 g/mol
Smeltepunkt: 286…288 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00278833
CAS-nummer: 15723-90-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR