5-(3-Chlorophenyl)-4-oxazolecarboxylic acid 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR59364-1G OR59364-5G
APOSOR59364-1GEA 1090 DKK
APOSOR59364-1G APOSOR59364-5G
5-(3-Chlorophenyl)-4-oxazolecarboxylic acid 98%
5-(3-Chlorophenyl)-4-oxazolecarboxylic acid
Formel: C₁₀H₆ClNO₃
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01934492
CAS-nummer: 255876-54-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR