(1-Methylpiperidin-4-yl)methanamine

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 1-methyl-piperidin-4-methylamine, (1-Methyl-4-piperidinyl)methanamine, (1-methylpiperidin-4-yl)methanamine

OR5892-1G OR5892-5G
APOSOR5892-1GEA 164 DKK
APOSOR5892-1G APOSOR5892-5G
(1-Methylpiperidin-4-yl)methanamine
(1-Methylpiperidin-4-yl)methanamine
Formel: C₇H₁₆N₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD05022430
CAS-nummer: 7149-42-0
EINECS: 230-477-2
UN nummer: 2735
ADR: 8,II

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR