2,4,6-Tribromtoluen 98%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
1-Methyl-2,4,6-tribromobenzene, 1,3,5-Tribromo-2-methylbenzene
OR5732-100G
OR5732-25G
OR5732-5G
APOSOR5732-100GEA
1400
DKK
APOSOR5732-100G
APOSOR5732-25G
APOSOR5732-5G
2,4,6-Tribromtoluen 98%
2,4,6-Tribromtoluen
Formel:
C₇H₅Br₃ Kogepunkt: 290 °C Smeltepunkt: 67…70 °C Massefylde: 2,479 g/cm³ (25 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00013527 CAS-nummer: 6320-40-7 EINECS: 228-672-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...