3-Bromo-N-isopropylbenzenesulphonamide 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

OR5727-1G OR5727-25G OR5727-5G
APOSOR5727-1GEA 806 DKK
APOSOR5727-1G APOSOR5727-25G APOSOR5727-5G
3-Bromo-N-isopropylbenzenesulphonamide 98%
3-Bromo-N-isopropylbenzenesulphonamide
Formel: C₉H₁₂BrNO₂S
Smeltepunkt: 52…56 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD07363820
CAS-nummer: 871269-08-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR