4-Chloropicolinamide 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR5724-100G OR5724-25G OR5724-5G OR5724-1G
APOSOR5724-100GEA 2850 DKK
APOSOR5724-100G APOSOR5724-25G APOSOR5724-5G APOSOR5724-1G
4-Chloropicolinamide 98%
4-Chloropicolinamide
Formel: C₆H₅ClN₂O
Smeltepunkt: 160…163 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01085339
CAS-nummer: 99586-65-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR