4-Bromo-N-isopropylbenzenesulphonamide 98%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
OR5695-1G
OR5695-5G
APOSOR5695-1GEA
230
DKK
APOSOR5695-1G
APOSOR5695-5G
4-Bromo-N-isopropylbenzenesulphonamide 98%
4-Bromo-N-isopropylbenzenesulphonamide
Formel:
C₉H₁₂BrNO₂S Molekylvægt: 278.17 g/mol Smeltepunkt: 87…89 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD01215090 CAS-nummer: 1984-27-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...