2-Brom-4-thiazolcarboxylsyre

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR5590-25G OR5590-5G
APOSOR5590-25GEA 4910 DKK
APOSOR5590-25G APOSOR5590-5G
2-Brom-4-thiazolcarboxylsyre
2-Brom-4-thiazolcarboxylsyre
Formel: C₄H₂BrNO₂S
Smeltepunkt: 212…214 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD04115729
CAS-nummer: 5198-88-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR