2-Amino-4-thiazolecarboxylic acid 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Aminothiazole-4-carboxylic acid

OR5587-25G OR5587-100G
APOSOR5587-25GEA 414 DKK
APOSOR5587-25G APOSOR5587-100G
2-Amino-4-thiazolecarboxylic acid 98%
2-Amino-4-thiazolecarboxylic acid
Formel: C₄H₄N₂O₂S
Smeltepunkt: 245…247 °C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00859429
CAS-nummer: 40283-41-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR