6-Acetylaminonicotinic acid 95%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR5583-1G
OR5583-250MG
OR5583-5G
APOSOR5583-1GEA
349
DKK
APOSOR5583-1G
APOSOR5583-250MG
APOSOR5583-5G
6-Acetylaminonicotinic acid 95%
6-Acetylaminonicotinic acid
Formel:
C₈H₈N₂O₃ Smeltepunkt: 276…278 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00129118 CAS-nummer: 21550-48-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...