2-Hydroxythiophenol 98%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
o-Hydroxythiophenol, 2-Hydroxyphenyl mercaptan, 2-Hydroxybenzene-1-thiol, 1-Hydroxy-2-mercaptobenzene, 2-Mercaptophenol, 2-Hydroxybenzenethiol
OR5576-25G
OR5576-5G
APOSOR5576-25GEA
3170
DKK
APOSOR5576-25G
APOSOR5576-5G
2-Hydroxythiophenol 98%
2-Hydroxythiophenol
Formel:
C₆H₆OS Kogepunkt: 217 °C Smeltepunkt: 19 °C Massefylde: 1,22 g/cm³ (25 °C) Flammepunkt: 97 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00040447 CAS-nummer: 1121-24-0 EINECS: 214-326-8 UN nummer: 2922 ADR: 8+6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...