Pentan-3-ylhydrazine dihydrochloride

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

OR55003-1G OR55003-5G
APOSOR55003-1GEA 1320 DKK
APOSOR55003-1G APOSOR55003-5G
Pentan-3-ylhydrazine dihydrochloride
Pentan-3-ylhydrazine dihydrochloride
Formel: C₅H₁₄N₂
Smeltepunkt: 60…80 °C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD12401615
CAS-nummer: 1211764-55-4
UN nummer: 2811
ADR: 6.1,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR