(R)-(S)-Josiphos 95%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR54583-1G
OR54583-250MG
APOSOR54583-1GEA
959
DKK
APOSOR54583-1G
APOSOR54583-250MG
(R)-(S)-Josiphos 95%
(R)-(S)-Josiphos
Formel:
C₃₆H₄₄FeP₂ Smeltepunkt: 95 °C (dec.) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00800284 CAS-nummer: 155806-35-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...