Benzyl(dimethyl)amin 99%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: DMBA, N,N-Dimethylbenzylamine, N-Benzyldimethylamine, BDMA

OR54497-2.5L OR54497-500ML
APOSOR54497-2.5LEA 1040 DKK
APOSOR54497-2.5L APOSOR54497-500ML
Benzyl(dimethyl)amin 99%
Benzyl(dimethyl)amin
Formel: C₆H₅CH₂N(CH₃)₂
Molekylvægt: 135.21 g/mol
Kogepunkt: 183…184 °C/0.75mmHg
Smeltepunkt: –75 °C
Massefylde: 0.9 g/cm³ (25 °C)
Flammepunkt: 53 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008329
CAS-nummer: 103-83-3
UN nummer: 2619
ADR: 8+3 II

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR