8-Aminoquinolin 98%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Quinolin-8-ylamine
OR5298-100G
OR5298-25G
OR5298-5G
APOSOR5298-100GEA
1370
DKK
APOSOR5298-100G
APOSOR5298-25G
APOSOR5298-5G
8-Aminoquinolin 98%
8-Aminoquinolin
Formel:
C₉H₈N₂ Kogepunkt: 174 °C/26mmHg Smeltepunkt: 66…68 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00006809 CAS-nummer: 578-66-5 EINECS: 209-427-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...