(-)-Diisopropyl-L-malate
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR52619-25G
OR52619-5G
APOSOR52619-25GEA
4310
DKK
APOSOR52619-25G
APOSOR52619-5G
(-)-Diisopropyl-L-malate
(-)-Diisopropyl-L-malate
Formel:
C₁₀H₁₈O₅ Kogepunkt: 236 °C Massefylde: 1,055 g/cm³ (25 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00274300 CAS-nummer: 83541-68-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...