L-Alanine benzyl ester hydrochloride 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR52412-100G OR52412-500G
APOSOR52412-100GEA 226 DKK
APOSOR52412-100G APOSOR52412-500G
L-Alanine benzyl ester hydrochloride 97%
L-Alanine benzyl ester hydrochloride
Formel: C₁₀H₁₃NO₂
Opbevaringstemperatur: Fryser
MDL Number: MFCD00054340
CAS-nummer: 5557-83-5
EINECS: 226-920-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR