2-Phenyl-1,3-propandiol 97%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
2-Phenylpropane-1,3-diol
OR52224-25G
OR52224-100G
APOSOR52224-25GEA
414
DKK
APOSOR52224-25G
APOSOR52224-100G
2-Phenyl-1,3-propandiol 97%
2-Phenyl-1,3-propandiol
Formel:
HOH₂CCH(C₆H₅)CH₂OH Smeltepunkt: 53…56 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00236056 CAS-nummer: 1570-95-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...