(S)-(+)-α-Methoxyphenyleddikesyre 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: (S)-(+)-α-Methoxyphenylacetic acid, (S)-alpha-Methoxyphenylacetic Acid

OR52197-1G OR52197-5G
APOSOR52197-1GEA 164 DKK
APOSOR52197-1G APOSOR52197-5G
(S)-(+)-α-Methoxyphenyleddikesyre 95%
(S)-(+)-α-Methoxyphenyleddikesyre
Formel: C₉H₁₀O₃
Molekylvægt: 166.18 g/mol
Kogepunkt: 164…166 °C/18mmHg
Smeltepunkt: 64…66 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00064216
CAS-nummer: 26164-26-1
EINECS: 247-492-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR