Disodium-4,4'-bis(2-sulphonatostyryl)biphenyl

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Sodium 2,2'-([1,1'-biphenyl]-4,4'-diylbis(ethene-2,1-diyl))dibenzenesulphonate, 4,4'-Bis(2-sulphonatostyryl)biphenyl Disodium Salt

OR52138-100G OR52138-500G OR52138-1KG
APOSOR52138-100GEA 175 DKK
APOSOR52138-100G APOSOR52138-500G APOSOR52138-1KG
Disodium-4,4'-bis(2-sulphonatostyryl)biphenyl
Disodium-4,4'-bis(2-sulphonatostyryl)biphenyl

Optical brightener.

Formel: C₂₈H₂₂O₆S₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00318874
CAS-nummer: 27344-41-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR