3-Chlor-4-nitrobenzoesyre
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR5193-1G
OR5193-25G
OR5193-5G
APOSOR5193-1GEA
164
DKK
APOSOR5193-1G
APOSOR5193-25G
APOSOR5193-5G
3-Chlor-4-nitrobenzoesyre
3-Chlor-4-nitrobenzoesyre
Formel:
C₇H₄ClNO₄ Smeltepunkt: 178…180 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD01632172 CAS-nummer: 39608-47-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...