4-Methoxystilbene 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR5076-25G OR5076-5G
APOSOR5076-25GEA 3750 DKK
APOSOR5076-25G APOSOR5076-5G
4-Methoxystilbene 98%
4-Methoxystilbene
Formel: C₁₅H₁₄O
Smeltepunkt: 134…136 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00017178
CAS-nummer: 1142-15-0
EINECS: 214-530-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR