Indol-3-carbaldehyd 99%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
1H-Indole-3-carbaldehyde, Indole-3-carboxaldehyde
OR5052-100G
OR5052-250G
OR5052-500G
APOSOR5052-100GEA
262
DKK
APOSOR5052-100G
APOSOR5052-250G
APOSOR5052-500G
Indol-3-carbaldehyd 99%
Indol-3-carbaldehyd
Formel:
C₉H₇NO Molekylvægt: 145.16 g/mol Smeltepunkt: 190…199 °C Flammepunkt: 240 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00005622 CAS-nummer: 487-89-8 EINECS: 207-665-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...