4-Acetylpyridin 97%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Methyl 4-pyridyl ketone
OR5047-25G
OR5047-100G
OR5047-1G
APOSOR5047-25GEA
469
DKK
APOSOR5047-25G
APOSOR5047-100G
APOSOR5047-1G
4-Acetylpyridin 97%
4-Acetylpyridin
Formel:
C₇H₇NO Kogepunkt: 210…212 °C Smeltepunkt: 13…16 °C Massefylde: 1,095 g/cm³ (25 °C) Flammepunkt: 104 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00006433 CAS-nummer: 1122-54-9 EINECS: 214-350-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...