Piperonylalkohol 98%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Benzo[d][1,3]dioxol-5-ylmethanol, 3,4-(Methylenedioxy)benzyl alcohol, 1,3-Benzodioxol-5-ylmethanol
OR5017-25G
OR5017-5G
APOSOR5017-25GEA
186
DKK
APOSOR5017-25G
APOSOR5017-5G
Piperonylalkohol 98%
Piperonylalkohol
Formel:
C₈H₈O₃ Kogepunkt: 156…158 °C Smeltepunkt: 50…55 °C Flammepunkt: >113 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00005836 CAS-nummer: 495-76-1 EINECS: 207-808-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...