2,3,4-Trimethoxybenzaldehyd 97%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
OR4984-250G
OR4984-50G
OR4984-1KG
APOSOR4984-250GEA
622
DKK
APOSOR4984-250G
APOSOR4984-50G
APOSOR4984-1KG
2,3,4-Trimethoxybenzaldehyd 97%
2,3,4-Trimethoxybenzaldehyde
Formel:
C₁₀H₁₂O₄ Kogepunkt: 168…170 °C/12mmHg Smeltepunkt: 38…42 °C Flammepunkt: >110 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00003310 CAS-nummer: 2103-57-3 EINECS: 218-271-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...