2-Amino-6-chloro-3-pyridinecarboxamide 95%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
OR480810-10G
OR480810-5G
OR480810-1G
APOSOR480810-10GEA
3820
DKK
APOSOR480810-10G
APOSOR480810-5G
APOSOR480810-1G
2-Amino-6-chloro-3-pyridinecarboxamide 95%
2-Amino-6-chloro-3-pyridinecarboxamide
Formel:
C₆H₆ClN₃O Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD12024435 CAS-nummer: 64321-24-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...