(Nitromethyl)benzene 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

OR480613-1G OR480613-25G OR480613-5G
APOSOR480613-1GEA 404 DKK
APOSOR480613-1G APOSOR480613-25G APOSOR480613-5G
(Nitromethyl)benzene 95%
(Nitromethyl)benzene
Formel: C₇H₇NO₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00053440
CAS-nummer: 622-42-4
UN nummer: 2810
ADR: 6.1,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR