3-Amino-4-bromo-6-chloropyridazine 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 4-Bromo-6-chloro-pyridazin-3-amine

OR480266-10G OR480266-25G OR480266-5G OR480266-1G
APOSOR480266-10GEA 0 DKK
APOSOR480266-10G APOSOR480266-25G APOSOR480266-5G APOSOR480266-1G
3-Amino-4-bromo-6-chloropyridazine 98%
3-Amino-4-bromo-6-chloropyridazine
Formel: C₄H₃BrClN₃
Molekylvægt: 208.4437 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD11520891
CAS-nummer: 446273-59-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR