(2-Chloro-4-methoxyphenyl)acetonitrile 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2-(2-Chloro-4-methoxyphenyl)acetonitrile

OR480252-10G OR480252-1G OR480252-5G
APOSOR480252-10GEA 7860 DKK
APOSOR480252-10G APOSOR480252-1G APOSOR480252-5G
(2-Chloro-4-methoxyphenyl)acetonitrile 97%
(2-Chloro-4-methoxyphenyl)acetonitrile
Formel: C₉H₈ClNO
Smeltepunkt: 41…43 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD08445710
CAS-nummer: 170737-93-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR