Fmoc-Cys(tBu)-OH 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: N-Fmoc-S-(tert-butyl)-L-cysteine, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-S-(tert-butyl)-L-cysteine, N-Fmoc-S-tert-butyl-L-cysteine, Fmoc-Cys(tBu)-OH

OR471634-100G OR471634-25G
APOSOR471634-100GEA 1570 DKK
APOSOR471634-100G APOSOR471634-25G
Fmoc-Cys(tBu)-OH 98%
Fmoc-Cys(tBu)-OH
Formel: C₂₂H₂₅NO₄S
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00037130
CAS-nummer: 67436-13-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR