2'-iodacetophenon
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
1-(2-Iodophenyl)ethan-1-one, 1-(2-Iodophenyl)ethanone, o-Iodoacetophenone
OR4681-100G
OR4681-25G
OR4681-5G
APOSOR4681-100GEA
1700
DKK
APOSOR4681-100G
APOSOR4681-25G
APOSOR4681-5G
2'-iodacetophenon
2'-iodacetophenon
Formel:
C₈H₇IO Kogepunkt: 139…140 °C/12mmHg Massefylde: 1,72 g/cm³ (25 °C) Flammepunkt: 103 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00094998 CAS-nummer: 2142-70-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...