3-Bromo-5-chloroanthranilic acid 97%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
2-Amino-3-bromo-5-chlorobenzoic acid
OR46070-1G
OR46070-5G
OR46070-25G
OR46070-10G
APOSOR46070-1GEA
284
DKK
APOSOR46070-1G
APOSOR46070-5G
APOSOR46070-25G
APOSOR46070-10G
3-Bromo-5-chloroanthranilic acid 97%
3-Bromo-5-chloroanthranilic acid
Formel:
C₇H₅BrClNO₂ Smeltepunkt: 225…229 °C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD04035658 CAS-nummer: 41198-02-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...