p-Toluenesulphonyl cyanide 97%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
4-Methylbenzenesulphonyl cyanide, Tosyl cyanide
OR46010-1G
OR46010-5G
OR46010-25G
APOSOR46010-1GEA
218
DKK
APOSOR46010-1G
APOSOR46010-5G
APOSOR46010-25G
p-Toluenesulphonyl cyanide 97%
p-Toluenesulphonyl cyanide
Formel:
C₈H₇NO₂S Molekylvægt: 181.21 g/mol Kogepunkt: 105…106 °C Smeltepunkt: 47…50 °C Flammepunkt: 110 °C Opbevaringstemperatur: Fryser |
MDL Number:
MFCD00009977 CAS-nummer: 19158-51-1 EINECS: 242-849-1 UN nummer: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...