3-Brom-4-methoxybenzoesyre

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR4560-25G OR4560-5G OR4560-100G
APOSOR4560-25GEA 317 DKK
APOSOR4560-25G APOSOR4560-5G APOSOR4560-100G
3-Brom-4-methoxybenzoesyre
3-Brom-4-methoxybenzoesyre
Formel: C₈H₇BrO₃
Smeltepunkt: 220…222 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00020295
CAS-nummer: 99-58-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR