2,4-Dimethylbenzylalkohol 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: (2,4-Dimethylphenyl)methanol, 4-(Hydroxymethyl)-m-xylene

OR451325-25G OR451325-5G
APOSOR451325-25GEA 806 DKK
APOSOR451325-25G APOSOR451325-5G
2,4-Dimethylbenzylalkohol 97%
2,4-Dimethylbenzylalkohol
Formel: C₉H₁₂O
Kogepunkt: 120 °C
Smeltepunkt: 25 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00004624
CAS-nummer: 16308-92-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR