Chroman-6-carbaldehyde

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

OR45040-1G OR45040-250MG
APOSOR45040-1GEA 2300 DKK
APOSOR45040-1G APOSOR45040-250MG
Chroman-6-carbaldehyde
Chroman-6-carbaldehyde
Formel: C₁₀H₁₀O₂
Kogepunkt: 98…104 °C/0,6mmHg
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD08146589
CAS-nummer: 55745-97-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR